Thermo Scientific Chemicals

Dilauroyl peroxide, 99%, Thermo Scientific Chemicals

Catalog number: 349745000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Dilauroyl peroxide, 99%, Thermo Scientific Chemicals

Catalog number: 349745000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 349745000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
105-74-8
IUPAC Name
dodecanoyl dodecaneperoxoate
Molecular Formula
C24H46O4
InChI Key
YIVJZNGAASQVEM-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC
Appearance (Color)
White
Appearance (Form)
Powder or granules or flakes or pellets
Infrared spectrum
Conforms
Titration Iodometric
>=98.5 %
Melting point
53°C to 57°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text