Thermo Scientific Chemicals

tert-Butyl peroxyacetate, 50% solution in aromatic free mineral spirit, Thermo Scientific Chemicals

Catalog number: 349862500
250 mL, Plastic bottle, Each
Thermo Scientific Chemicals

tert-Butyl peroxyacetate, 50% solution in aromatic free mineral spirit, Thermo Scientific Chemicals

Catalog number: 349862500
250 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 349862500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-71-1
IUPAC Name
tert-butyl ethaneperoxoate
Molecular Formula
C6H12O3
InChI Key
SWAXTRYEYUTSAP-UHFFFAOYSA-N
SMILES
CC(=O)OOC(C)(C)C
Appearance (Form)
Clear solution
Color scale
=<20 APHA
Titration Iodometric
49 to 51 %
Impurity
=<1000 ppm Hydroperoxides (as TBHP)
Hydrolyzable chloride
=<50 ppm Inorganic + organic

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text