1x1 image pixel for data collection
Di-tert-butyl peroxide, 99%
Di-tert-butyl peroxide, 99%
Di-tert-butyl peroxide, 99%
Di-tert-butyl peroxide, 99%
Thermo Scientific Chemicals

Di-tert-butyl peroxide, 99%

CAS: 110-05-4 | C8H18O2 | 146.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
250 g
Packaging:
Plastic bottle
Catalog number 349931000
Price (USD)
38.65
Online Exclusive
42.70
Save 4.05 (9%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.70
Save 4.05 (9%)
Each
Add to cart
Chemical Identifiers
CAS110-05-4
IUPAC Name2-(tert-butylperoxy)-2-methylpropane
Molecular FormulaC8H18O2
InChI KeyLSXWFXONGKSEMY-UHFFFAOYSA-N
SMILESCC(C)(C)OOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Hydroperoxides=<0.1 % (as TBHP)
Hydroperoxides=<0.1 % (as TBHP)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link