Di-tert-butyl peroxide, 99%, Thermo Scientific Chemicals
Di-tert-butyl peroxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-tert-butyl peroxide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
100 g
Packaging:
Plastic bottle
Catalog number 349932500
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
73.40 
Save 10.75 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
73.40 
Save 10.75 (15%)
Add to cart
Di-tert-butyl peroxide, 99%, Thermo Scientific Chemicals
Catalog number349932500
Price (USD)/ Each
62.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
73.40 
Save 10.75 (15%)
-
Add to cart
Chemical Identifiers
CAS110-05-4
IUPAC Name2-(tert-butylperoxy)-2-methylpropane
Molecular FormulaC8H18O2
InChI KeyLSXWFXONGKSEMY-UHFFFAOYSA-N
SMILESCC(C)(C)OOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3880 to 1.3900 (20°C, 589 nm)
GC=<0.1 % Tert-butyl hydroperoxide
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only