3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number 349941000
Price (USD)/ Each
70.50
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
70.50
Add to cart
3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, Thermo Scientific Chemicals
Catalog number349941000
Price (USD)/ Each
70.50
-
Add to cart
Chemical Identifiers
IUPAC Name3,6,9-triethyl-3,6,9-trimethyl-1,2,4,5,7,8-hexaoxonane
Molecular FormulaC12H24O6
InChI KeyKVWLLOIEGKLBPA-UHFFFAOYSA-N
SMILESCCC1(C)OOC(C)(CC)OOC(C)(CC)OO1
Molecular Weight (g/mol)264.32
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Color scale=<50 APHA (at 40°C)
Titration Iodometric7.3 to 7.6 % (total active oxygen)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only