Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 349960050
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.00 
Save 5.35 (13%)
-
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.00 
Save 5.35 (13%)
Add to cart
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Catalog number349960050
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.00 
Save 5.35 (13%)
-
Add to cart
Chemical Identifiers
CAS80-15-9
IUPAC Name2-phenylpropane-2-peroxol
Molecular FormulaC9H12O2
InChI KeyYQHLDYVWEZKEOX-UHFFFAOYSA-N
SMILESCC(C)(OO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<450 APHA
Infrared spectrumConforms
Titration Iodometric80 to 85 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only