Tributyl(vinyl)tin, 97%, Thermo Scientific Chemicals
Tributyl(vinyl)tin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tributyl(vinyl)tin, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 350000250
Price (USD)/ Each
368.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
434.00 
Save 65.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
368.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
434.00 
Save 65.35 (15%)
Add to cart
Tributyl(vinyl)tin, 97%, Thermo Scientific Chemicals
Catalog number350000250
Price (USD)/ Each
368.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
434.00 
Save 65.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7486-35-3
IUPAC Nametributyl(ethenyl)stannane
Molecular FormulaC14H30Sn
InChI KeyQIWRFOJWQSSRJZ-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4770 to 1.4790 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only