6-Chloro-2,4-difluoroaniline, 97%, Thermo Scientific Chemicals
6-Chloro-2,4-difluoroaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Chloro-2,4-difluoroaniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 350200010
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
96.90 
Save 14.25 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
96.90 
Save 14.25 (15%)
Add to cart
6-Chloro-2,4-difluoroaniline, 97%, Thermo Scientific Chemicals
Catalog number350200010
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
96.90 
Save 14.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-chloro-4,6-difluoroaniline
Molecular FormulaC6H4ClF2N
InChI KeyDUPRZIYJXCCXQZ-UHFFFAOYSA-N
SMILESNC1=C(F)C=C(F)C=C1Cl
Molecular Weight (g/mol)163.55
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Low melting solid
Melting point28°C to 33°C
Appearance (Color)Off-white to grey-green
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only