Thermo Scientific Chemicals

chloro(diethyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: 350310050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

chloro(diethyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: 350310050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 350310050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
686-69-1
IUPAC Name
chlorodiethylphosphane
Molecular Formula
C4H10ClP
InChI Key
INJBDKCHQWVDGT-UHFFFAOYSA-N
SMILES
CCP(Cl)CC
Appearance (Form)
Liquid
NMR
>=94 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4740 to 1.4760 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text