2,3-Diamino-5-chloropyridine, 97%, Thermo Scientific Chemicals
2,3-Diamino-5-chloropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Diamino-5-chloropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 350340050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25710-20-7
IUPAC Name2,3-diamino-5-chloropyridin-1-ium
Molecular FormulaC5H7ClN3
InChI KeyZRCMCGQDIYNWDX-UHFFFAOYSA-O
SMILESNC1=CC(Cl)=C[NH+]=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 %
Infrared spectrumConforms
Appearance (Color)White to grey to brown
Appearance (Form)Powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only