GlutaMAX™ Supplement
Try new Pen-Strep antibiotic with GlutaMAX supplement, a stable alternative to L-glutamine that enhances cell health. Learn more >
GlutaMAX™ Supplement
Gibco™

GlutaMAX™ Supplement

Green features
Gibco GlutaMAX Supplement is an alternative to L-glutamine, with increased stability that improves cell health. GlutaMAX Supplement is suitable forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
3505007920 x 100 mL
35050061100 mL
Catalog number 35050079
Price (USD)/ Each
892.00
-
Add to cart
Quantity:
20 x 100 mL
Price (USD)/ Each
892.00
Add to cart
GlutaMAX™ Supplement
Catalog number35050079
Price (USD)/ Each
892.00
-
Add to cart

Gibco GlutaMAX Supplement is an alternative to L-glutamine, with increased stability that improves cell health. GlutaMAX Supplement is suitable for both adherent and suspension culture of mammalian cells, with no adaptation required. GlutaMAX Supplement is offered as a 200 mM L-alanyl-L-glutamine dipeptide in 0.85% NaCl. GlutaMAX Supplement is also included in a variety of media formulations. Compared to L-glutamine, GlutaMAX Supplement:

• Minimizes toxic ammonia build-up
• Improves cell viability and growth
• Remains stable across a wide range of temperatures

Minimizes toxic ammonia build-up
Unlike L-glutamine, GlutaMAX Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.

Improves cell viability and growth
Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX Supplement that is not possible with L-glutamine.

Remains stable across a wide range of temperatures
GlutaMAX Supplement is room-temperature stable and ready to use. GlutaMAX Supplement retains full functionality for 24 months at room temperature, 2 to 8°C, or -5 to -30°C for convenient storage and handling. GlutaMAX Supplement is even stable at 37°C for a week or more.

Specifications
Cell TypeMammalian Cells
ClassificationAnimal Origin-free
Concentration100 X
Culture TypeMammalian Cell Culture
FormLiquid
Product TypeGlutaMax
Shelf Life24 Months
SterilitySterile-filtered
For Use With (Application)Clinical Research, Translational Research
Green FeaturesSustainable packaging
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality440 - 500 mOsm/kg
pH4.7 to 6
Product LineGibco™, GlutaMAX™
Quantity20 x 100 mL
SourceAnimal origin-free
Unit SizeEach
Contents & Storage
Storage conditions: 15°C to 30°C
Alternate storage conditions: -5°C to -20°C and 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture