2-Bromobenzylamine, 98%, Thermo Scientific Chemicals
2-Bromobenzylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromobenzylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 350510250
Price (USD)/ Each
272.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
272.00
Add to cart
2-Bromobenzylamine, 98%, Thermo Scientific Chemicals
Catalog number350510250
Price (USD)/ Each
272.00
-
Add to cart
Chemical Identifiers
CAS3959-05-5
IUPAC Name1-(2-bromophenyl)methanamine
Molecular FormulaC7H8BrN
InChI KeyNOYASZMZIBFFNZ-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.5880 to 1.5930 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only