2-Bromophenylacetone, 99%, Thermo Scientific Chemicals
2-Bromophenylacetone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromophenylacetone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 350590010
Price (USD)/ Each
38.60
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
38.60
Add to cart
2-Bromophenylacetone, 99%, Thermo Scientific Chemicals
Catalog number350590010
Price (USD)/ Each
38.60
-
Add to cart
Chemical Identifiers
CAS21906-31-0
IUPAC Name1-(2-bromophenyl)propan-2-one
Molecular FormulaC9H9BrO
InChI KeyTZIAZLUAMDLDJF-UHFFFAOYSA-N
SMILESCC(=O)CC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5575 to 1.5595 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only