Thermo Scientific Chemicals

Triethylamine trihydrofluoride, ca. 37% HF, Thermo Scientific Chemicals

Catalog number: 350621000
100 mL, Plastic bottle, Each
Thermo Scientific Chemicals

Triethylamine trihydrofluoride, ca. 37% HF, Thermo Scientific Chemicals

Catalog number: 350621000
100 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 350621000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
73602-61-6
IUPAC Name
trihydrogen triethylamine trifluoride
Molecular Formula
C6H18F3N
InChI Key
IKGLACJFEHSFNN-UHFFFAOYSA-N
SMILES
[H+].[H+].[H+].[F-].[F-].[F-].CCN(CC)CC
Appearance (Color)
Clear colorless to yellow to brown
Titration with NaOH
21.3 to 28.0 % (Available HF)
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Titration
32.0 to 42.0 % (Calculated Total HF)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text