3-Chlorophenylacetone, 94%, Thermo Scientific Chemicals
3-Chlorophenylacetone, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chlorophenylacetone, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
View all
Glass Bottle
Catalog number 350690250
Price (USD)/ Each
284.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
284.00
Add to cart
3-Chlorophenylacetone, 94%, Thermo Scientific Chemicals
Catalog number350690250
Price (USD)/ Each
284.00
-
Add to cart
Chemical Identifiers
IUPAC Name1-(3-chlorophenyl)propan-2-one
Molecular FormulaC9H9ClO
InChI KeyVCNYPJMEQHTAHS-UHFFFAOYSA-N
SMILESCC(=O)CC1=CC=CC(Cl)=C1
Molecular Weight (g/mol)168.62
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=92 %
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.5320 to 1.5360 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only