Pentaethylene glycol, 98%, Thermo Scientific Chemicals
Pentaethylene glycol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentaethylene glycol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 350730050
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
90.50 
Save 13.85 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
90.50 
Save 13.85 (15%)
Add to cart
Pentaethylene glycol, 98%, Thermo Scientific Chemicals
Catalog number350730050
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
90.50 
Save 13.85 (15%)
-
Add to cart
Chemical Identifiers
CAS4792-15-8
IUPAC Name3,6,9,12-tetraoxatetradecane-1,14-diol
Molecular FormulaC10H22O6
InChI KeyJLFNLZLINWHATN-UHFFFAOYSA-N
SMILESOCCOCCOCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4610 to 1.4630 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only