3,4-Dichlorophenethyl alcohol, 97%, Thermo Scientific Chemicals
3,4-Dichlorophenethyl alcohol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dichlorophenethyl alcohol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 350770010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(3,4-dichlorophenyl)ethan-1-ol
Molecular FormulaC8H8Cl2O
InChI KeyGITOMJDYNUMCOV-UHFFFAOYSA-N
SMILESOCCC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol)191.05
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Refractive index1.5620 to 1.5680 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only