Thermo Scientific Chemicals

Methyl 2-chlorobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 351311000
100 g, Each
Thermo Scientific Chemicals

Methyl 2-chlorobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 351311000
100 g, Each
Quantity
Catalog number: 351311000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
610-96-8
IUPAC Name
methyl 2-chlorobenzoate
Molecular Formula
C8H7ClO2
InChI Key
JAVRNIFMYIJXIE-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1Cl
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.5350 to 1.5370 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text