Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, Thermo Scientific Chemicals

Catalog number: 351421000
100 mL, Each
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, Thermo Scientific Chemicals

Catalog number: 351421000
100 mL, Each
Quantity
Catalog number: 351421000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
865-49-6
IUPAC Name
trichloro(²H)methane
Molecular Formula
CHCl3
InChI Key
HEDRZPFGACZZDS-MICDWDOJSA-N
SMILES
[2H]C(Cl)(Cl)Cl
Water
=<0.02 % (Coulometric)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
D-Enrichment
>=99.75 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text