Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, Thermo Scientific Chemicals
Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 351425000
Price (USD)/ Each
373.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
439.00 
Save 65.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
373.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
439.00 
Save 65.35 (15%)
Add to cart
Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, Thermo Scientific Chemicals
Catalog number351425000
Price (USD)/ Each
373.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
439.00 
Save 65.35 (15%)
-
Add to cart
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.75 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only