Thermo Scientific Chemicals

Deuterium oxide, for NMR, packaged in 0.75ml ampoules, 99.8 atom% D, Thermo Scientific Chemicals

Catalog number: 351430075
7.5 mL, Each
Thermo Scientific Chemicals

Deuterium oxide, for NMR, packaged in 0.75ml ampoules, 99.8 atom% D, Thermo Scientific Chemicals

Catalog number: 351430075
7.5 mL, Each
Quantity
Catalog number: 351430075
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-20-0
IUPAC Name
[(²H)oxy](²H)
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES
[2H]O[2H]
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
D-Enrichment
>=99.75 %
Tritium (T)
=<370 Bq/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text