Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D, Thermo Scientific Chemicals
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
7.5 mL
Packaging:
Ampule
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 351480075
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
180.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
180.00 
Save 26.35 (15%)
Add to cart
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D, Thermo Scientific Chemicals
Catalog number351480075
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
180.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(²H₅)pyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-RALIUCGRSA-N
SMILES[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Molecular Weight (g/mol)84.13
View more
SpecificationsSpecification SheetSpecification Sheet
D-Enrichment>=99.75 %
Water=<0.03 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only