Benzyl 2,2,2-trichloroacetimidate, 99%, ACROS Organics™
Benzyl 2,2,2-trichloroacetimidate, 99%, ACROS Organics™
Thermo Scientific Chemicals

Benzyl 2,2,2-trichloroacetimidate, 99%, ACROS Organics™

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 351601000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS81927-55-1
IUPAC Namebenzyl 2,2,2-trichloroethanimidate
Molecular FormulaC9H8Cl3NO
InChI KeyHUZCTWYDQIQZPM-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C(=N)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5425 to 1.5465 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only