3-Aminopropionitrile, 98%, stabilized, Thermo Scientific Chemicals
3-Aminopropionitrile, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminopropionitrile, 98%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 351750250
Price (USD)/ Each
357.65
Online exclusive
420.00 
Save 62.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
357.65
Online exclusive
420.00 
Save 62.35 (15%)
Add to cart
3-Aminopropionitrile, 98%, stabilized, Thermo Scientific Chemicals
Catalog number351750250
Price (USD)/ Each
357.65
Online exclusive
420.00 
Save 62.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name3-aminopropanenitrile
Molecular FormulaC3H6N2
InChI KeyAGSPXMVUFBBBMO-UHFFFAOYSA-N
SMILESNCCC#N
Molecular Weight (g/mol)70.10
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizerpotassium carbonate
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only