Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 351760050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 351760050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 351760050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4919-37-3
IUPAC Name
4-hydroxy-3,5-dimethylbenzoate
Molecular Formula
C9H9O3
InChI Key
OMNHTTWQSSUZHO-UHFFFAOYSA-M
SMILES
CC1=CC(=CC(C)=C1O)C([O-])=O
Appearance (Color)
Off-white to light beige to light pink
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Melting point
221°C to 226°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text