4,4'-Dibromobenzil, 97%, Thermo Scientific Chemicals
4,4'-Dibromobenzil, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Dibromobenzil, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 351780100
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.10 
Save 9.45 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.10 
Save 9.45 (15%)
Add to cart
4,4'-Dibromobenzil, 97%, Thermo Scientific Chemicals
Catalog number351780100
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.10 
Save 9.45 (15%)
-
Add to cart
Chemical Identifiers
CAS35578-47-3
IUPAC Namebis(4-bromophenyl)ethane-1,2-dione
Molecular FormulaC14H8Br2O2
InChI KeyNYCBYBDDECLFPE-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)Yellow
Melting point223°C to 228°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only