Thermo Scientific Chemicals

Isosorbide, 98%, Thermo Scientific Chemicals

Catalog number: 351915000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Isosorbide, 98%, Thermo Scientific Chemicals

Catalog number: 351915000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 351915000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
652-67-5
IUPAC Name
(3R,3aR,6S,6aR)-hexahydrofuro[3,2-b]furan-3,6-diol
Molecular Formula
C6H10O4
InChI Key
KLDXJTOLSGUMSJ-JGWLITMVSA-N
SMILES
O[C@H]1CO[C@@H]2[C@H](O)CO[C@H]12
Appearance (Color)
Off-white to yellow to beige
Appearance (Form)
Crystals or crystalline solid or mass
Infrared spectrum
Conforms
Melting point
56°C to 62°C
Specific optical rotation
+42° to +48° (20°C, 589 nm) (c=3, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text