N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals
N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 351940500
Price (USD)/ Each
52.50
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
52.50
Add to cart
N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals
Catalog number351940500
Price (USD)/ Each
52.50
-
Add to cart
Chemical Identifiers
CAS5036-48-6
IUPAC Name3-(1H-imidazol-1-yl)propan-1-amine
Molecular FormulaC6H11N3
InChI KeyKDHWOCLBMVSZPG-UHFFFAOYSA-N
SMILESNCCCN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5170 to 1.5210 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only