Thermo Scientific Chemicals

Methyl 3-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 352001000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 352001000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 352001000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19438-10-9
IUPAC Name
methyl 3-hydroxybenzoate
Molecular Formula
C8H8O3
InChI Key
YKUCHDXIBAQWSF-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(O)=C1
Appearance (Color)
White to beige
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
70°C to 74°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text