Tetra-n-octylammonium bromide, 98%, Thermo Scientific Chemicals
Tetra-n-octylammonium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-octylammonium bromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 352100050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14866-33-2
IUPAC Nametetraoctylazanium bromide
Molecular FormulaC32H68BrN
InChI KeyQBVXKDJEZKEASM-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Powder or flakes
Infrared spectrumConforms
Titration Argentometric>=97.5 %
Melting point95°C to 99°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only