Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals
Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 352120250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1193-82-4
IUPAC Namemethanesulfinylbenzene
Molecular FormulaC7H8OS
InChI KeyJXTGICXCHWMCPM-UHFFFAOYNA-N
SMILESCS(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid or fused mass
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.5755 to 1.5795 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only