mono-Methyl terephthalate, 97%, Thermo Scientific Chemicals
mono-Methyl terephthalate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

mono-Methyl terephthalate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 352341000
Price (USD)/ Each
445.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
445.00
Add to cart
mono-Methyl terephthalate, 97%, Thermo Scientific Chemicals
Catalog number352341000
Price (USD)/ Each
445.00
-
Add to cart
Chemical Identifiers
IUPAC Name4-(methoxycarbonyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyREIDAMBAPLIATC-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol)180.16
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Melting point219°C to 224°C
Appearance (Color)White to off-white
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only