Thermo Scientific Chemicals

1,2,4,5-Tetrabromobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 352400250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2,4,5-Tetrabromobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 352400250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 352400250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
636-28-2
IUPAC Name
1,2,4,5-tetrabromobenzene
Molecular Formula
C6H2Br4
InChI Key
QCKHVNQHBOGZER-UHFFFAOYSA-N
SMILES
BrC1=CC(Br)=C(Br)C=C1Br
Infrared spectrum
Conforms
Melting point
177°C to 183°C
GC
>=96.0 %
Appearance (Form)
Powder or crystals or needles
Appearance (Color)
White to beige to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text