Thermo Scientific Chemicals

L-Ascorbic acid sodium salt, 99%

Catalog number: 352681000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

L-Ascorbic acid sodium salt, 99%

Catalog number: 352681000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 352681000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
134-03-2
IUPAC Name
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
Molecular Formula
C6H7NaO6
InChI Key
IFVCRSPJFHGFCG-HXPAKLQESA-N
SMILES
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration Iodimetric
98.5 to 101.5 %
Solubility
(5% in water) Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text