4-Phenoxybenzoic acid, 98%, Thermo Scientific Chemicals
4-Phenoxybenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenoxybenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 352720250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-phenoxybenzoic acid
Molecular FormulaC13H10O3
InChI KeyRYAQFHLUEMJOMF-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Molecular Weight (g/mol)214.22
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Melting point160°C to 164°C
Infrared spectrumConforms
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only