Thermo Scientific Chemicals

Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 352940010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 352940010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 352940010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2855-13-2
IUPAC Name
3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
Molecular Formula
C10H22N2
InChI Key
RNLHGQLZWXBQNY-UHFFFAOYNA-N
SMILES
CC1(C)CC(N)CC(C)(CN)C1
Infrared spectrum
Conforms
Appearance (Form)
Clear liquid
Refractive index
1.4880 to 1.4900 (20°C, 589 nm)
GC
>=99.0 % (Sum of isomers)
Water
=<0.5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text