Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number 353091000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS429-42-5
IUPAC Nametetrabutylazanium; tetrafluoroboranuide
Molecular FormulaC16H36BF4N
InChI KeyNNZZSJSQYOFZAM-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=97.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only