Thermo Scientific Chemicals

Tetrakis(dimethylamino)titanium, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 353560250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrakis(dimethylamino)titanium, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 353560250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 353560250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3275-24-9
IUPAC Name
titanium(4+) tetrakis(dimethylazanide)
Molecular Formula
C8H24N4Ti
InChI Key
MNWRORMXBIWXCI-UHFFFAOYSA-N
SMILES
[Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C
Appearance (Color)
Clear yellow to orange
Total trace metal impurities
=<0.01 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text