Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 354081000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 354081000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 354081000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65039-09-0
IUPAC Name
1-ethyl-3-methyl-1H-imidazol-3-ium chloride
Molecular Formula
C6H11ClN2
InChI Key
BMQZYMYBQZGEEY-UHFFFAOYSA-M
SMILES
[Cl-].CCN1C=C[N+](C)=C1
Appearance (Color)
White to yellow to orange
Appearance (Form)
Adhering powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text