Thermo Scientific Chemicals

1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Catalog number: 354211000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals

Catalog number: 354211000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 354211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
174501-65-6
IUPAC Name
1-butyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular Formula
C8H15BF4N2
InChI Key
LSBXQLQATZTAPE-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1
Appearance (Color)
Clear yellow to orange
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
HPLC
>=98.0 %
Water
=<1.0 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text