Thermo Scientific Chemicals

Methanephosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: 354331000
100 g, Each
Thermo Scientific Chemicals

Methanephosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: 354331000
100 g, Each
Quantity
Catalog number: 354331000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
993-13-5
IUPAC Name
methylphosphonic acid
Molecular Formula
CH5O3P
InChI Key
YACKEPLHDIMKIO-UHFFFAOYSA-N
SMILES
CP(O)(O)=O
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Melting point
103°C to 107°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text