Cyclopentanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Cyclopentanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentanecarbonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 354370050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4524-93-0
IUPAC Namecyclopentanecarbonyl chloride
Molecular FormulaC6H9ClO
InChI KeyWEPUZBYKXNKSDH-UHFFFAOYSA-N
SMILESClC(=O)C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Titration RCOCl>=97.5 %
Refractive index1.4610 to 1.4640 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only