Thermo Scientific Chemicals

3,4-Dimethoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 354390250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dimethoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 354390250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 354390250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23095-31-0
IUPAC Name
3,4-dimethoxybenzene-1-sulfonyl chloride
Molecular Formula
C8H9ClO4S
InChI Key
RSJSYCZYQNJQPY-UHFFFAOYSA-N
SMILES
COC1=C(OC)C=C(C=C1)S(Cl)(=O)=O
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Melting point
65°C to 70°C
HPLC
>=96.0 %
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text