Thermo Scientific Chemicals

TentaGel™ MB-Br, Thermo Scientific Chemicals

Catalog number: 354480010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

TentaGel™ MB-Br, Thermo Scientific Chemicals

Catalog number: 354480010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 354480010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
73342-17-3
Appearance (Color)
White to yellow
Loading
0.4 to 0.6 mmol/g
Particle size
140 to 170 µm
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text