TentaGel™ S-NH2, Thermo Scientific Chemicals
TentaGel™ S-NH2, Thermo Scientific Chemicals
Thermo Scientific Chemicals
TentaGel™ S-NH2, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 354610050
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
Add to cart
TentaGel™ S-NH2, Thermo Scientific Chemicals
Catalog number354610050
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialTentaGel™ S-NH2
ColorWhite to Yellow
Physical FormPowder
Loading0.2 to 0.3mmol/g
PackagingGlass bottle
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only