Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 354800025
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
Add to cart
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
Catalog number354800025
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.005 %
Stabilizer0.003 to 0.006 % (Amylene)
Residue after evaporation=<0.0005 %
GC>=99.9 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For peptide synthesis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only