2,7-Dibromofluorene, 99%, Thermo Scientific Chemicals
2,7-Dibromofluorene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,7-Dibromofluorene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 355240250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2,7-dibromo-9H-fluorene
Molecular FormulaC13H8Br2
InChI KeyAVXFJPFSWLMKSG-UHFFFAOYSA-N
SMILESBrC1=CC=C2C(CC3=CC(Br)=CC=C23)=C1
Molecular Weight (g/mol)324.02
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point164°C to 167°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only