Potassium superoxide
Potassium superoxide
Potassium superoxide
Potassium superoxide
Thermo Scientific Chemicals

Potassium superoxide

CAS: 12030-88-5 | KO2 | 71.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 355425000
Price (EUR)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS12030-88-5
IUPAC Namepotassium dioxygen
Molecular FormulaKO2
InChI KeyCLSKHAYBTFRDOV-UHFFFAOYSA-N
SMILES[K+].O=O
View more
SpecificationsSpecification SheetSpecification Sheet
Oxygen (O)>=215 cm³/g O2-content
Appearance (Form)Powder or chunks
Appearance (Color)Yellow
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Oxidizing solids
Category 2
Skin Corrosion/Irritation
Category 1
Serious Eye Damage/Eye Irritation
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H272 - May intensify fire; oxidizer

H314 - Causes severe skin burns and eye damage

EU Specific Hazard Statements

EUH014 - Reacts violently with water

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P220 - Keep away from clothing and other combustible materials

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

P371 + P380 + P375 - In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion

Share catalog number, name or link

1x1 image pixel for data collection