Thermo Scientific Chemicals

2-Amino-5-iodobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 355440250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Amino-5-iodobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 355440250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 355440250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5326-47-6
IUPAC Name
2-amino-5-iodobenzoic acid
Molecular Formula
C7H6INO2
InChI Key
GOLGILSVWFKZRQ-UHFFFAOYSA-N
SMILES
NC1=CC=C(I)C=C1C(O)=O
Titration with TBAH
>=97.5 %
Appearance (Color)
Beige to grey to purple
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text