Benchtop Coolers
Benchtop Coolers
Thermo Scientific™

Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature RangeArrayHolds
355501-20°C4 x 520 Tubes (0.2 to 0.5 mL)
5115-0012-20°C3 x 412 Tubes (0.2 to 0.5 mL)
5115-0032-20°C4 x 832 Tubes (0.2 to 0.5 mL)
5116-13000°C3 x 412 Tubes (12 to 13 mm dia.)
5116-16000°C3 x 412 Tubes (16 to 17 mm dia.)
DS5116-00120°C3 x 412 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
DS5116-00320°C4 x 832 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
DS5114-0012-20°C3 x 412 Cryogenic Vials (0.2 to 1.5 mL)
Catalog number 355501
Price (USD)
250.20
Online Exclusive
292.00
Save 41.80 (14%)
Each
-
Add to cart
Temperature Range:
-20°C
Array:
4 x 5
Holds:
20 Tubes (0.2 to 0.5 mL)
Price (USD)
250.20
Online Exclusive
292.00
Save 41.80 (14%)
Each
Add to cart

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications
Array4 x 5
Description4x5 compartments, -20 deg.
DesignNon-filled/clear lid
Dimensions (L x W x H)8.94 x 6.5 x 8.25 in. (226 x 165 x 133 mm)
Height (English)8.25 in.
Height (Metric)133 mm
Holds Temp., Hours<-15°C, 1
Length (English)8.94 in.
Length (Metric)226 mm
Width (English)6.5 in.
Width (Metric)165 mm
Capacity (Metric)0.2 to 0.5 mL
ColorClear
Holds20 Tubes (0.2 to 0.5 mL)
MaterialPolycarbonate
Temperature Range-20°C
TypeBenchtop Cooler
Unit SizeEach
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link